Order Entry
Austria
Orders LinkContactUsLinkComponent
Face shield, Tuffmaster™

Face shield, Tuffmaster™

Anti-UV, anti-scratch, PC, 230×370 mm
  111-1635
 :   3M
Face shield, Tuffmaster™
  111-1635
 :  3M
Face shield, Tuffmaster™
Face shield, Tuffmaster™

Anti-UV, anti-scratch, PC, 230×370 mm

 
53,60 €
   
1 * 1 ST
 

This face shield consists of headgear (H8) made of thermoplastic and clear PC visor (WP96). Both parts provide high impact resistance. Ideal for use in metalworking, manufacturing, construction, mining and transportation.

  • Adjustable ratchet suspension of the headgear allows visor to be replaced in seconds
  • Ratchet helps to provide a precise fit and comfortable head protection
  • Five position crown strap for additional support and comfort
  • Moulded PC visor provides impact and splash resistance and mounts quickly to any 3M™ headgear without tools, snaps or cams

Visor thickness: 2 mm

Certifications: EN 166

Documents

 
  • Type:
    Anti-UV, anti-scratch
  • L×W:
    230×370 mm
  • Material:
    PC
Accessories

 

  •  
    Description
     
     
  • 111-1634
    Visor, clear moulded PC, 2 mm thickness
       
    1 * 1 ST - 9,20 €

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.