Order Entry
Austria
ContactUsLinkComponent
DL-α-Tocopherol (Vitamin E), dry powder sprayed, EMPROVE® ESSENTIAL Ph. Eur., BP, SAFC®
DL-α-Tocopherol (Vitamin E), dry powder sprayed, EMPROVE® ESSENTIAL Ph. Eur., BP, SAFC®
Catalog # 5.00878.9024
CAS Number:  
DL-α-Tocopherol (Vitamin E), dry powder sprayed, EMPROVE® ESSENTIAL Ph. Eur., BP, SAFC®
Catalog # 5.00878.9024
CAS Number:  

Specifications

  • Pk:
    20 kg
  • Seq:
    4241

Specifications

Specification Test Results

  • all-rac-α-Tocopheryl acetate content (GC, Ph Eur):
    50.0 - 57.5 %
  • Identity (GC):
    passes test
  • As (Arsenic):
    ≤ 0.0003 %
  • Cu (Copper):
    ≤ 50 ppm
  • Pb (Lead):
    ≤ 10 ppm
  • Zn (Zinc):
    ≤ 25 ppm
  • Ethanol (HS-GC):
    ≤ 5000 ppm
  • Acetic acid (HS-GC):
    ≤ 5000 ppm
  • Heptane (HS-GC):
    ≤ 5000 ppm
  • Methanol (HS-GC):
    ≤ 3000 ppm
  • Pyridine (HS-GC):
    ≤ 200 ppm
  • Toluene (HS-GC):
    ≤ 890 ppm
  • Residual solvents (ICH Q3C):
    excluded by production process
  • Loss on drying:
    ≤ 5.0 %
  • Total aerobic microbial count (TAMC):
    ≤ 1000 CFU/g
  • Total combined yeasts/moulds count (TYMC):
    ≤ 100 CFU/g
  • Bile-tolerant gram-negative bacteria (absent in 1 g):
    passes test
  • Candida albicans (absent in 1 g):
    passes test
  • Escherichia coli (absent in 1 g):
    passes test
  • Pseudomonas aeruginosa (absent in 1 g):
    passes test
  • Salmonella (absent in 10 g):
    passes test
  • Staphylococcus aureus (absent in 1 g):
    passes test

Specification Test Results

About this item

 :
 :430.71 g/mol
EINECS:233-466-0
CAS Number:10191-41-0
 :1406-66-2
Formula:C₂₉H₅₀O₂
MDL Number:MFCD00072051