ContactUsLinkComponent
VWR®, Multi-Use Collection and Storage Tray
VWR®, Multi-Use Collection and Storage Tray
Catalog # 216-1492
Supplier:  VWR Collection
undefined
VWR®, Multi-Use Collection and Storage Tray
Catalog # 216-1492
Supplier:  VWR Collection
Specifications
  • W×D×H:
    480×331×84 mm
  • Pk:
    1
Specifications
About this item
Plastic, blue tray with white insert.
  • Designed to hold a large number of samples, whilst being easy to carry
  • For storage on an average size refrigerator shelf
  • Hard wearing and flexible
  • Large integrated label holder for clear and easy identification
  • Can be cleaned with common disinfectants and germicides
For clinical samples. Great for transporting and storing general laboratory items. Lightweight and sturdy. Can hold up to 24 full specimen containers up to 58 mm diameter, using the insert. Removable insert to allow outer shell to be used for storage or transportation of laboratory products. Stackable with other trays to support as many as four high.