Reagents for water hardness, Testomat®
Supplier: Heyl GmbH & Co, Gebr.
Certificates
About this item
Frequently Bought Together
Potassium chloride 3 mol/l (3 N) in aqueous solution
Supplier: WTW
Methanol ≥99.8%, HiPerSolv CHROMANORM® Reag. Ph. Eur., gradient grade for HPLC
Supplier: VWR Chemicals
Acetone ≥99%, TECHNICAL
Supplier: VWR Chemicals
Test kit, water hardness, DUROVAL® B
Supplier: Heyl GmbH & Co, Gebr.
Measuring beakers with handle
Supplier: VITLAB
VWR® Dosatest, pH indicator test strips, non bleeding
Supplier: VWR Chemicals
VWR® High Performance, Bottles, Wide Neck with Screw Cap
Supplier: VWR Collection
VWR®, Nitrile Gloves
Supplier: VWR Collection
Disinfectants, ClearKlens DE
Supplier: Diversey
pH indicator strips, MQuant®, Supelco®
Supplier: Merck
Mixing measuring cylinders, tall form, class B, soda-lime glass
Supplier: GLASWARENFABRIK KARL HECHT
[EN]CONTAINER ALUMINIUM 96X78X24MM 125 M 1 * 100 ST
Supplier: Novelis
Caprylic acid 99%
Supplier: Thermo Fisher Scientific
Reagents for water hardness, Testomat®
Supplier: Heyl GmbH & Co, Gebr.