ContactUsLinkComponent
Volumetric flasks, class A, glass, clear, NS plastic stopper
Volumetric flasks, class A, glass, clear, NS plastic stopper
Catalog # WITG3.664.0030
Supplier:  witeg Labortechnik
undefined
Volumetric flasks, class A, glass, clear, NS plastic stopper
Catalog # WITG3.664.0030
Supplier:  witeg Labortechnik
Specifications
  • NS:
    10/19
  • Capacity:
    30 ml
  • Pk:
    1
  • Tolerance:
    0,04 ± ml
Specifications
About this item
DURAN®, borosilicate glass, clear, with PE stopper.
  • With acid and alkali resistant ring marks and inscriptions
  • DIFFICO stain markings
Certifications: DIN 12664