Micro-volume spectrophotometer, Thermo Scientific™ NanoDrop™ Lite
732-2980
- Model:ND-Lite
- Range:260 / 280 nm
- Absorbance range:0 − 30 A
- Detector:Silicon photodiode
- Light source:LED
- Sample size:1 - 2 µl
- Description:Micro-volume spectrophotometer
- W×D×H:160×115×95 mm
- Absorbance accuracy:±3 %
- Concentration:4 − 1500 ng/l (dsDNA) ; 3 − 1200 ng/l (RNA) ; 0,12 − 45 mg/ml Protein (BSA)
- Concentration units:4 − 1500 ng/l (dsDNA) ; 3 − 1200 ng/l (RNA) ; 0,12 − 45 mg/ml Protein (BSA)
- Interfaces:USB
- Optical design:Filter
- Photometric accuracy:3 % (1,5 A @ 260 nm)
- Photometric range:0 - 30 Abs (10 mm equivalent)
- Spectral bandwidth:8 nm
- Wavelength range:260 (nucleic acid concentration) / 280 (purified protein concentration)
- Pk:1
The NanoDrop Lite is designed for high precision measurements of the concentration of DNA, RNA and proteins on the microvolume scale.
- Employs the NanoDrop microvolume sampling technology
- Extremely compact with built-in controls − Software without computer required
- Optional dockable label printer available for samples and laboratory records
- Intuitive software for concentration determinations
- Smaller footprint due to integration of the control unit