Orders LinkContactUsLinkComponent
Corning® Lymphocyte Separation Medium, Corning
Supplier:   Corning
Corning® Lymphocyte Separation Medium, Corning
Supplier:  Corning
Certificates
Corning® Lymphocyte Separation Medium, Corning
Corning® Lymphocyte Separation Medium, Corning
2 Options Available Below
Product Details & Documents
Lymphocyte Separation Medium (LSM) was originally designed for the in vitro isolation of lymphocytes from diluted whole blood
It is a sterile, iso-osmotic polysucrose and diatrizoate solution with low viscosity and a density of 1.077-1.080 g/mL at 20ºC.
Product Family Options
Product Information
  • Availability
    Price
  • MDTC25-072-CI
    LSM (Lymphocyte Separation Medium), 1×100 ml
    LSM (Lymphocyte Separation Medium), 1×100 ml
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 52,68 €
    52,68 €
    1 * 1 ST
       52,68 €
  • 25-072-CV
    LSM (Lymphocyte Separation Medium), 1×500 ml
    LSM (Lymphocyte Separation Medium), 1×500 ml
    Log in for availability
    Log in for availability
    1 * 500 mL
    1 * 500 mL - 154,68 €
    154,68 €
    1 * 500 mL
       154,68 €