Order Entry
Austria
ContactUsLinkComponent
Glove box, fibreglass, HEPA filtered, Protector®
Glove box, fibreglass, HEPA filtered, Protector®
Supplier:  LABCONCO
Certificates
undefined
Glove box, fibreglass, HEPA filtered, Protector®
Supplier:  LABCONCO

Specifications

  • Catalog No:
  • LBCP5065031
  • LBCP5065035
  • Model:
  • Protector®
  • Protector®
  • Connection:
  • 208 - 230 V, 10 amps, 50/60 Hz
  • 208 - 230 V, 10 amps, 50/60 Hz
  • Observation window (W×H):
  • 914,5×701 mm
  • 914,5×701 mm
  • W×D×H ext.:
  • 1623×787×1161 mm
  • 1623×787×1161 mm
  • Weight:
  • 227 kg
  • 227 kg

Specifications

Related Information

Frequently Bought Together