Anti-c-Myc tag Mouse Monoclonal Antibody (DyLight® 550) [clone: 9E10]
COBSD9-1714
- Antibody type:Tag
- Antigen name:v-myc avian myelocytomatosis viral oncogene homolog
- Antigen symbol:MYC
- Clonality:Monoclonal
- Clone:9E10
- Conjugation:DyLight® 550
- ELISA:Yes
- Flow cytometry:Yes
- Host:Mouse
- ImmunoChemistry:Yes
- Isotype:IgG1 kappa
- Reactivity:
- Western blot:Yes
- Environmentally Preferable:
- Cross adsorption:none
- Gene ID:4609
- Antigen synonyms:MRTL|bHLHe39|c-Myc|MYCC
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µg
- Pk:100 µG
Type: Tag
Antigen: c-Myc tag (c-Myc tag)
Clonality: monoclonal
Clone: 90000000000
Conjugation: DyLight® 550
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: