Order Entry
Austria
ContactUsLinkComponent
Anti-6X His tag Mouse Monoclonal Antibody (DyLight® 488) [clone: AD1.1.10]
Anti-6X His tag Mouse Monoclonal Antibody (DyLight® 488) [clone: AD1.1.10]
Catalog # COBSD8-1711
Anti-6X His tag Mouse Monoclonal Antibody (DyLight® 488) [clone: AD1.1.10]
Catalog # COBSD8-1711

Specifications

  • Antibody type:
    Tag
  • Antigen name:
    egl-9 family hypoxia-inducible factor 2
  • Antigen symbol:
    EGLN2
  • Clonality:
    Monoclonal
  • Clone:
    AD1.1.10
  • Conjugation:
    DyLight® 488
  • ELISA:
    Yes
  • Flow cytometry:
    Yes
  • Host:
    Mouse
  • ImmunoChemistry:
    Yes
  • Isotype:
    IgG1 kappa
  • Reactivity:
  • Western blot:
    Yes
  • Environmentally Preferable:
  • Cross adsorption:
    none
  • Gene ID:
    112398
  • Antigen synonyms:
    HIFPH1|HIF-PH1|EIT6|HPH-1|HPH-3|PHD1
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Size:
    100 µg
  • Pk:
    100 µG

Specifications

About this item

Type: Tag
Antigen: 6X His tag (6X His tag)
Clonality: monoclonal
Clone: AD1.1.10
Conjugation: DyLight® 488
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: