Anti-PGIS Rabbit Polyclonal Antibody RPE (R-Phycoerythrin)
Artikelnr. COBSD5-1867
Lieferant: Columbia Biosciences
Spezifikationen
- Antibody type:Primary
- Antigen symbol:PGIS
- Clonality:Polyclonal
- Conjugation:RPE (R-Phycoerythrin)
- ELISA:Yes
- Flow cytometry:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:
- Environmentally Preferable:
- Cross adsorption:none
- Amino acid number:475 to 490
- Sequence:PEFDLSRYGFGLMQPE
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µg
- Pk:100 µG
Spezifikationen
Über diesen Artikel
Type: Primary
Antigen: PGIS sequence amino acids 475-490 (PEFDLSRYGFGLMQPE)
Clonality: polyclonal
Clone:
Conjugation: RPE (R-Phycoerythrin)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: