Order Entry
Austria
ContactUsLinkComponent
Anti-EP4 receptor C-terminal amino acids 459-488 (GSGRAGPAPKGSSLQVTFPSETLNLSEKCI) Rabbit Polyclonal Antibody (APC (Allophycocyanin))
Anti-EP4 receptor C-terminal amino acids 459-488 (GSGRAGPAPKGSSLQVTFPSETLNLSEKCI) Rabbit Polyclonal Antibody (APC (Allophycocyanin))
  COBSD3-1866-50
Anti-EP4 receptor C-terminal amino acids 459-488 (GSGRAGPAPKGSSLQVTFPSETLNLSEKCI) Rabbit Polyclonal Antibody (APC (Allophycocyanin))
  COBSD3-1866-50

 

  • Antibody type:
    Primary
  • Antigen name:
    EP4 receptor
  • Clonality:
    Polyclonal
  • Conjugation:
    APC (Allophycocyanin)
  • ELISA:
    Yes
  • Flow cytometry:
    Yes
  • Host:
    Rabbit
  • Isotype:
    IgG
  • Reactivity:
  • Environmentally Preferable:
  • Epitope:
    C-terminal
  • Cross adsorption:
    none
  • Amino acid number:
    459 to 488
  • Sequence:
    GSGRAGPAPKGSSLQVTFPSETLNLSEKCI
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Size:
    100 µg
  • Pk:
    50 µG

 

 

Type: Primary
Antigen: EP4 receptor C-terminal amino acids 459-488 (GSGRAGPAPKGSSLQVTFPSETLNLSEKCI)
Clonality: polyclonal
Clone:
Conjugation: APC (Allophycocyanin)
Epitope: C-terminal
Host: Rabbit
Isotype: IgG
Reactivity: