Mouse IgG2b Isotype Control (APC (Allophycocyanin))
100 µg
Catalog # COBSD3-1602B
Supplier: Columbia Biosciences
Mouse IgG2b Isotype Control (APC (Allophycocyanin))
Catalog # COBSD3-1602B
Supplier: Columbia Biosciences
Jump to:
Product Details & Documents
Type: Control
Antigen: IgG2b (Mouse) (mouse IgG2b)
Clonality: polyclonal
Clone:
Conjugation: APC (Allophycocyanin)
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: None
Documents
Specifications
- Size:100 µg
- Cat. No.:COBSD3-1602B
- Clonality:Polyclonal
- Flow cytometry:Yes
- Antibody type:Control
- Isotype:IgG2b
- Storage temperature:Cool
- Host:Mouse
- Conjugation:APC (Allophycocyanin)
- Cross adsorption:none
- Shipping temperature:Ice
- ELISA:Yes



