Anti-V5 Tag Rabbit Polyclonal Antibody (Peridinin Chlorophyll)
Catalog # COBSD11-1834
Supplier: Columbia Biosciences
Specifications
- Antibody type:Tag
- Antigen name:V5 tag
- Antigen symbol:V5
- Clonality:Polyclonal
- Conjugation:PerCP (Peridinin-Chlorophyll Protein Complex)
- ELISA:Yes
- Flow cytometry:Yes
- Host:Rabbit
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:
- Western blot:Yes
- Environmentally Preferable:
- Cross adsorption:none
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Type: Tag
Antigen: V5 Tag (V5 tag)
Clonality: polyclonal
Clone:
Conjugation: Peridinin Chlorophyll
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: