Anti-Ricinus communis RCA60, RCA120 Goat Polyclonal Antibody
USBIR2031-51C
- Antibody type:Primary
- Antigen symbol:Ricinus communis RCA60, RCA120
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Goat
- Isotype:IgG
- Form:lyophilized powder
- Storage buffer:10mMphosphate, pH 7.8, 0,15M sodium chloride, 0,08% sodium azide, sucrose has been for stabilization.
- Storage temperature:Cool
- Concentration:~2 mg/ml (after reconstitution)
- Shipping temperature:Ice
- Purification:purified by affinity chromatography
- Size:1 mg
- Pk:1 mg
Recommended dilution: ELISA: 1-10 ?g/ml
Type: Primary
Antigen: Ricinus communis RCA60, RCA120
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: