Anti-JAM-A, Domain 2 Rabbit Polyclonal Antibody
Catalog # USBIJ8009-05D
Supplier: US Biological
Specifications
- Antibody type:Primary
- Antigen name:JAM-A, Domain 2
- Antigen symbol:JAM-A
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Flow cytometry:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human
- Western blot:Yes
- Form:liquid
- Storage buffer:PBS, 0,1% BSA, 0,02% sodium azide.
- Storage temperature:Cool
- Concentration:~0,1 mg/ml
- Shipping temperature:Ice
- Purification:purified by Protein A chromatography
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Recommended dilution: Flow Cytometry: 1:50; Western Blot: 1:50
Type: Primary
Antigen: JAM-A, Domain 2
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human