Order Entry
Austria
ContactUsLinkComponent
Anti-Cyclic AMP Dependent Protein Kinase Regulatory Subunit 1b Sheep Polyclonal Antibody
Anti-Cyclic AMP Dependent Protein Kinase Regulatory Subunit 1b Sheep Polyclonal Antibody
Catalog # USBIC8450-09A
Supplier:  US Biological
Anti-Cyclic AMP Dependent Protein Kinase Regulatory Subunit 1b Sheep Polyclonal Antibody
Catalog # USBIC8450-09A
Supplier:  US Biological

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Cyclic AMP Dependent Protein Kinase Regulatory Subunit 1b
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • Host:
    Sheep
  • Isotype:
    IgG
  • Reactivity:
    Human,
    Rat,
    Mouse
  • Western blot:
    Yes
  • Form:
    lyophilized powder
  • Storage buffer:
    PBSwith 5% trehalose.
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Purification:
    purified by immunoaffinity chromatography
  • Size:
    100 µg
  • Pk:
    100 µG

Specifications

About this item

Type: Primary
Antigen: Cyclic AMP Dependent Protein Kinase Regulatory Subunit 1b
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Sheep
Isotype: IgG
Reactivity: Human, Mouse, Rat