Anti-DEFB103B Mouse monoclonal antibody unconjugated [clone: L3-18b-E1]
USBIB0899-03H-100
- Antibody type:Primary
- Antigen name:defensin, beta 103B
- Antigen symbol:DEFB103B
- Clonality:Monoclonal
- Clone:L3-18b-E1
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Isotype:IgG1
- Reactivity:Human
- Form:lyophilized powder
- Amino acid number:6 to 22
- Storage buffer:50mM Tris, pH 7,4.
- Sequence:LQKYYCRVRGGRCAVLS
- Storage temperature:Cool
- Concentration:~1 mg/ml
- Shipping temperature:Ice
- Tested applications:Radio Immunoassay
- Purification:purified by Protein G chromatography
- Size:100 µg
- Pk:100 µG
Type: Primary
Antigen: DEFB103B (defensin, beta 103B)
Clonality: monoclonal
Clone: L3-18b-E1
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human