Anti-VCAM1 Mouse Monoclonal Antibody [clone: 13j97]
USBI142757
- Antibody type:Primary
- Antigen name:Vascular cell adhesion molecule 1
- Antigen symbol:VCAM1
- Clonality:Monoclonal
- Clone:13j97
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- ImmunoChemistry:Yes
- Isotype:
- Reactivity:Human
- Western blot:Yes
- Environmentally Preferable:
- Form:liquid
- Storage buffer:PBS, pH 7,2, 0,05% sodium azide, 50% glycerol.
- Storage temperature:Cool
- Concentration:~0,5 mg/ml
- Shipping temperature:Ice
- Purification:purified by immunoaffinity chromatography
- Size:200 µg
- Pk:200 µG
Recommended dilution: ELISA: 1:100-200; Western Blot: 1:100-400; Immunochemistry: 1:100-500
Type: Primary
Antigen: VCAM1 (vascular cell adhesion molecule 1)
Clonality: monoclonal
Clone: 13j97
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human