Anti-MBTPS1 Mouse Monoclonal Antibody [clone: 2E6]
Catalog # USBI129473
Supplier: US Biological
Specifications
- Antibody type:Primary
- Antigen name:Membrane-bound transcription factor peptidase site 1
- Antigen symbol:MBTPS1
- Clonality:Monoclonal
- Clone:2E6
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- ImmunoFluorescence:Yes
- Isotype:IgG1 kappa
- Reactivity:Human
- Environmentally Preferable:
- Form:liquid
- Amino acid number:246 to 355
- Storage buffer:PBS, pH 7,2.
- Sequence:GLGHGTFVAGVIASMRECQGFAPDAELHIFRVFTNNQVSYTSWFLDAFNYAILKKIDVLNLSIGGPDFMDHPFVDKVWELTANNVIMVSAIGNDGPLYGTLNNPADQMDV
- Storage temperature:Cool
- Shipping temperature:Ice
- Purification:purified by Protein A chromatography
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Recommended dilution: Immunofluorescence: 10 ?g/ml
Type: Primary
Antigen: MBTPS1 (membrane-bound transcription factor peptidase, site 1)
Clonality: monoclonal
Clone: 2000000
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: Human