Anti-CSDE1 Mouse Polyclonal Antibody
USBI125387
- Antibody type:Primary
- Antigen name:cold shock domain containing E1, RNA-binding
- Antigen symbol:CSDE1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Mouse
- Isotype:IgG
- Reactivity:Human
- Western blot:Yes
- Form:liquid
- Amino acid number:1 to 767
- Storage buffer:PBS, pH 7,2.
- Storage temperature:Cool
- Shipping temperature:Ice
- Purification:purified by Protein A chromatography
- Size:50 µg
- Pk:50 µG
Type: Primary
Antigen: CSDE1 (cold shock domain containing E1, RNA-binding)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG
Reactivity: Human