Anti-ATP6V1E1 Mouse Monoclonal Antibody [clone: 4E11]
Catalog # USBI123734
Supplier: US Biological
Specifications
- Antibody type:Primary
- Antigen name:ATPase, H+ transporting, lysosomal 31kDa, V1 subunit E1
- Antigen symbol:ATP6V1E1
- Clonality:Monoclonal
- Clone:4E11
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Isotype:IgG2a kappa
- Reactivity:Human
- Western blot:Yes
- Form:liquid
- Storage buffer:PBS, pH 7,2.
- Sequence:MALSDADVQKQIKHMMAFIEQEANEKAEEIDAKAEEEFNIEKGRLVQTQRLKIMEYYEKKEKQIEQQKKIQMSNLMNQARLKVLRARDDLITDLLNEAKQRLSKVVKDTTRYQVLLDGLVLQGLYQLLEPRMIVRCRKQDFPLVKAAVQKAIPMYKIATKNDVDVQIDQESYLPEDIAGGVEIYNGDRKIKVSNTLESRLDLIAQQMMPEVRGALFGANANRKFLD
- Storage temperature:Cool
- Shipping temperature:Ice
- Purification:purified by Protein A chromatography
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Type: Primary
Antigen: ATP6V1E1 (ATPase, H+ transporting, lysosomal 31kDa, V1 subunit E1)
Clonality: monoclonal
Clone: 400000000000
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a Kappa
Reactivity: Human