Anti-PACS2 Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))
Catalog # USBI039605-HRP
Supplier: US Biological
Specifications
- Antibody type:Primary
- Antigen name:phosphofurin acidic cluster sorting Protein 2
- Antigen symbol:PACS2
- Clonality:Polyclonal
- Conjugation:HRP (Horseradish Peroxidase)
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human
- Western blot:Yes
- Epitope:C-terminal
- Form:liquid
- Storage buffer:PBS, pH 7,2
- Storage temperature:Cool
- Shipping temperature:Ice
- Purification:purified by Protein A chromatography
- Size:200 µl
- Pk:200 µl
Specifications
About this item
Type: Primary
Antigen: PACS2 (phosphofurin acidic cluster sorting protein 2)
Clonality: Polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope: C-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human