Order Entry
Austria
ContactUsLinkComponent
 
Anti-UB Mouse Monoclonal Antibody
  QEDB11019
Anti-UB Mouse Monoclonal Antibody
  QEDB11019

 

  • Antibody type:
    Primary
  • Antigen name:
    Ubiquitin
  • Antigen symbol:
    UB
  • Clonality:
    Monoclonal
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Mouse
  • Isotype:
    IgG2a
  • Reactivity:
    Human,
    Bovine,
    Rat,
    Mouse
  • Western blot:
    Yes
  • Cross adsorption:
    none
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Size:
    100 µg
  • Pk:
    100 µG

 

 

Type: Primary
Antigen: UB (ubiquitin)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Bovine, Human, Mouse, Rat