Order Entry
Austria
ContactUsLinkComponent
Anti-SYK Rabbit Polyclonal Antibody
Anti-SYK Rabbit Polyclonal Antibody
Catalog # PK1010-50
Anti-SYK Rabbit Polyclonal Antibody
Catalog # PK1010-50

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Spleen Tyrosine Kinase
  • Antigen symbol:
    SYK
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Rabbit
  • Isotype:
  • Reactivity:
  • Western blot:
    Yes
  • Environmentally Preferable:
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Size:
    50 µl
  • Pk:
    50 µl

Specifications

About this item

Type: Primary
Antigen: SYK (spleen tyrosine kinase)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: