Order Entry
Austria
ContactUsLinkComponent
Anti-p53 (Ab-6) (Pantropic) (DO-1) Mouse Monoclonal Antibody [clone: DO-1]
Anti-p53 (Ab-6) (Pantropic) (DO-1) Mouse Monoclonal Antibody [clone: DO-1]
Catalog # OP43-100
Anti-p53 (Ab-6) (Pantropic) (DO-1) Mouse Monoclonal Antibody [clone: DO-1]
Catalog # OP43-100

Specifications

  • Antibody type:
    Primary
  • Antigen symbol:
    P53
  • Clonality:
    Monoclonal
  • Clone:
    DO-1
  • Conjugation:
    Unconjugated
  • Host:
    Mouse
  • ImmunoChemistry:
    Yes
  • ImmunoFluorescence:
    Yes
  • ImmunoPrecipitation:
    Yes
  • Isotype:
  • Reactivity:
  • Western blot:
    Yes
  • Environmentally Preferable:
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Size:
    100 µg
  • Pk:
    100 µG

Specifications

About this item

Type: Primary
Antigen: p53 (Ab-6) (Pantropic) (DO-1)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity: