Anti-c-K-Ras (Ab-1) (234-4.2) Mouse Monoclonal Antibody [clone: 234-4.2]
Catalog # OP24-100
Supplier: Merck Millipore (Oncogene)
Specifications
- Antibody type:Primary
- Antigen symbol:c-K-Ras
- Clonality:Monoclonal
- Clone:234-4.2
- Conjugation:Unconjugated
- Host:Mouse
- ImmunoChemistry:Yes
- ImmunoPrecipitation:Yes
- Isotype:
- Reactivity:
- Western blot:Yes
- Environmentally Preferable:
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Type: Primary
Antigen: c-K-Ras (Ab-1) (234-4.2)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity: