Anti-c-ErbB2/c-Neu (Ab-3) (3B5) Mouse Monoclonal Antibody [clone: 3B5]
OP15-100
- Antibody type:Primary
- Antigen name:V-erb-b2 avian erythroblastic leukemia viral oncogene homolog 2
- Antigen symbol:C-ERBB2
- Clonality:Monoclonal
- Clone:3B5
- Conjugation:Unconjugated
- Host:Mouse
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- ImmunoPrecipitation:Yes
- Western blot:Yes
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µg
- Pk:100 µG
Type: Primary
Antigen: c-ErbB2/c-Neu (Ab-3) (3B5)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity: