Anti-MDM2 (Ab-5) (4B2C1.11) Mouse Monoclonal Antibody [clone: 4B2C1.11]
OP145-100
- Antibody type:Primary
- Antigen symbol:MDM2
- Clonality:Monoclonal
- Clone:4B2C1.11
- Conjugation:Unconjugated
- Host:Mouse
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- ImmunoPrecipitation:Yes
- Isotype:
- Reactivity:Human
- Western blot:Yes
- Environmentally Preferable:
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µg
- Pk:100 µG
Type: Primary
Antigen: MDM2 (Ab-5) (4B2C1.11)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity: Human