Anti-c-ErbB3 (Ab-4) (H3.90.6 or Ab90) Mouse Monoclonal Antibody [clone: H3.90.6 or Ab90]
Catalog # OP119-100
Supplier: Merck Millipore (Oncogene)
Specifications
- Antibody type:Primary
- Antigen name:V-erb-b2 avian erythroblastic leukemia viral oncogene homolog 3
- Antigen symbol:C-ERBB3
- Clonality:Monoclonal
- Clone:H3.90.6 or Ab90
- Conjugation:Unconjugated
- Flow cytometry:Yes
- Host:Mouse
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- ImmunoPrecipitation:Yes
- Isotype:
- Reactivity:Human
- Western blot:Yes
- Environmentally Preferable:
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Type: Primary
Antigen: c-ErbB3 (Ab-4) (H3.90.6 or Ab90)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity: Human