Anti-MMP-9 (Ab-1) (6-6B) Mouse Monoclonal Antibody [clone: 6-6B]
Catalog # IM09L-100
Supplier: Merck Millipore (Oncogene)
Specifications
- Antibody type:Primary
- Antigen symbol:MMP-9
- Clonality:Monoclonal
- Clone:6-6B
- Conjugation:Unconjugated
- Host:Mouse
- ImmunoPrecipitation:Yes
- Isotype:
- Reactivity:Human
- Western blot:Yes
- Environmentally Preferable:
- Neutralisation:Yes
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Type: Primary
Antigen: MMP-9 (Ab-1) (6-6B)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity: Human