Anti-Vinculin (V284 or VLN01) Mouse Monoclonal Antibody [clone: V284 or VLN01]
CP74-100
- Antibody type:Primary
- Antigen name:Vinculin
- Clonality:Monoclonal
- Clone:V284 or VLN01
- Conjugation:Unconjugated
- Host:Mouse
- ImmunoChemistry:Yes
- ImmunoPrecipitation:Yes
- Isotype:
- Reactivity:Human
- Western blot:Yes
- Environmentally Preferable:
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µg
- Pk:100 µG
Type: Primary
Antigen: Vinculin (V284 or VLN01)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity: Human