Order Entry
Austria
ContactUsLinkComponent
Anti-CDH1 Mouse Monoclonal Antibody [clone: DH01]
Anti-CDH1 Mouse Monoclonal Antibody [clone: DH01]
Catalog # CC43-100
Anti-CDH1 Mouse Monoclonal Antibody [clone: DH01]
Catalog # CC43-100

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    cadherin 1, type 1, E-cadherin (epithelial)
  • Antigen symbol:
    CDH1
  • Clonality:
    Monoclonal
  • Clone:
    DH01
  • Conjugation:
    Unconjugated
  • Host:
    Mouse
  • ImmunoPrecipitation:
    Yes
  • Isotype:
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Environmentally Preferable:
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Size:
    100 µg
  • Pk:
    100 µG

Specifications

About this item

Type: Primary
Antigen: CDH1 (cadherin 1, type 1, E-cadherin (epithelial))
Clonality: monoclonal
Clone: DH01
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity: Human