Anti-Tissue Factor (TF9-10H10) Mouse Monoclonal Antibody [clone: TF9-10H10]
612161-500
- Antibody type:Primary
- Antigen name:Tissue Factor
- Antigen symbol:TF
- Clonality:Monoclonal
- Clone:TF9-10H10
- Conjugation:Unconjugated
- ELISA:Yes
- Flow cytometry:Yes
- Host:Mouse
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Western blot:Yes
- Neutralisation:Yes
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:500 µg
- Pk:500 µG
Type: Primary
Antigen: Tissue Factor (TF9-10H10)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity: