Order Entry
Austria
ContactUsLinkComponent
 
Anti-PAI-1 (MA-33B8) Mouse Monoclonal Antibody [clone: MA-33B8]
  528215-100
Anti-PAI-1 (MA-33B8) Mouse Monoclonal Antibody [clone: MA-33B8]
  528215-100

 

  • Antibody type:
    Primary
  • Antigen symbol:
    PAI-1
  • Clonality:
    Monoclonal
  • Clone:
    MA-33B8
  • Conjugation:
    Unconjugated
  • Host:
    Mouse
  • Isotype:
  • Reactivity:
  • Western blot:
    Yes
  • Environmentally Preferable:
  • Neutralisation:
    Yes
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Size:
    100 µg
  • Pk:
    100 µG

 

 

Type: Primary
Antigen: PAI-1 (MA-33B8)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity: