Order Entry
Austria
ContactUsLinkComponent
Anti-Phosphotyrosine (PY20) Mouse Monoclonal Antibody [clone: PY20] Peroxidase
Anti-Phosphotyrosine (PY20) Mouse Monoclonal Antibody [clone: PY20] Peroxidase
Catalog # 525320-100
Anti-Phosphotyrosine (PY20) Mouse Monoclonal Antibody [clone: PY20] Peroxidase
Catalog # 525320-100

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Phosphotyrosine
  • Antigen symbol:
    PY
  • Clonality:
    Monoclonal
  • Clone:
    PY20
  • Conjugation:
    Peroxidase
  • ELISA:
    Yes
  • Host:
    Mouse
  • ImmunoChemistry:
    Yes
  • Reactivity:
    Multiple
  • Western blot:
    Yes
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Size:
    100 µg
  • Pk:
    100 µG

Specifications

About this item

Type: Primary
Antigen: Phosphotyrosine (PY20)
Clonality: monoclonal
Clone:
Conjugation: Peroxidase
Epitope:
Host: Mouse
Isotype: none
Reactivity: Pan