Order Entry
Austria
ContactUsLinkComponent
Anti-Nitric Oxide Synthase (1131-1144), Inducible Rabbit Polyclonal Antibody
Anti-Nitric Oxide Synthase (1131-1144), Inducible Rabbit Polyclonal Antibody
Catalog # 482728-100
Anti-Nitric Oxide Synthase (1131-1144), Inducible Rabbit Polyclonal Antibody
Catalog # 482728-100

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Nitric Oxide Synthase (1131-1144, Inducible)
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • ImmunoFluorescence:
    Yes
  • Isotype:
  • Reactivity:
  • Western blot:
    Yes
  • Environmentally Preferable:
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Size:
    100 µl
  • Pk:
    100 µl

Specifications

About this item

Type: Primary
Antigen: Nitric Oxide Synthase (1131-1144), Inducible
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: