Order Entry
Austria
ContactUsLinkComponent
 
Anti-Fodrin (Fod009) Mouse Monoclonal Antibody
  344050-50
Anti-Fodrin (Fod009) Mouse Monoclonal Antibody
  344050-50

 

  • Antibody type:
    Primary
  • Antigen name:
    Fodrin
  • Antigen symbol:
    Fod009
  • Clonality:
    Monoclonal
  • Conjugation:
    Unconjugated
  • Flow cytometry:
    Yes
  • Host:
    Mouse
  • ImmunoFluorescence:
    Yes
  • Isotype:
  • Reactivity:
  • Western blot:
    Yes
  • Environmentally Preferable:
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Size:
    50 µg
  • Pk:
    50 µG

 

 

Type: Primary
Antigen: Fodrin (Fod009)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity: