Anti-CD18 (IB4) Mouse Monoclonal Antibody
217660-100
- Antibody type:Primary
- Antigen symbol:CD18
- Clonality:Monoclonal
- Conjugation:Unconjugated
- Flow cytometry:Yes
- Host:Mouse
- ImmunoChemistry:Yes
- ImmunoPrecipitation:Yes
- Isotype:
- Reactivity:Human
- Environmentally Preferable:
- Antigen synonyms:IB4
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µg
- Pk:100 µG
Type: Primary
Antigen: CD18 (IB4)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity: Human