Anti-CALR Mouse Monoclonal Antibody [clone: FMC75]
Catalog # 208912-50
Supplier: Merck Millipore (Oncogene)
Specifications
- Antibody type:Primary
- Antigen name:calreticulin
- Antigen symbol:CALR
- Clonality:Monoclonal
- Clone:FMC75
- Conjugation:Unconjugated
- Flow cytometry:Yes
- Host:Mouse
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:
- Reactivity:
- Western blot:Yes
- Environmentally Preferable:
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:50 µg
- Pk:50 µG
Specifications
About this item
Type: Primary
Antigen: CALR (calreticulin)
Clonality: monoclonal
Clone: FMC75
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity: