Anti-AQP5 Rabbit Polyclonal Antibody
Catalog # 178615-50
Supplier: Merck Millipore (Oncogene)
Specifications
- Antibody type:Primary
- Antigen name:Aquaporin 5
- Antigen symbol:AQP5
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:
- Reactivity:
- Western blot:Yes
- Environmentally Preferable:
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:50 µg
- Pk:50 µG
Specifications
About this item
Type: Primary
Antigen: AQP5 (aquaporin 5)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: