Anti-HSA Mouse Monoclonal Antibody [clone: 2C7B4]
Catalog # GENSA01731-40
Supplier: Genscript
Specifications
- Antibody type:Primary
- Antigen symbol:HSA
- Clonality:Monoclonal
- Clone:2C7B4
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Isotype:IgG1 kappa
- Reactivity:
- Western blot:Yes
- Environmentally Preferable:
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:40 µg
- Pk:40 µG
Specifications
About this item
Type: Primary
Antigen: HSA
Clonality: monoclonal
Clone: 2C7B4
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: