Order Entry
Austria
ContactUsLinkComponent
 
Anti-Cre Recombinase Mouse monoclonal antibody unconjugated
  GENSA00888-40
 :  Genscript
undefined
Anti-Cre Recombinase Mouse monoclonal antibody unconjugated
  GENSA00888-40
 :  Genscript

 

  • Antibody type:
    Tag
  • Clonality:
    Monoclonal
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Mouse
  • Isotype:
    IgG2a
  • Western blot:
    Yes
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Size:
    40 µg
  • Pk:
    40 µG

 

 

Type: Tag
Antigen: Cre Recombinase
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a (Mouse)
Reactivity: