Order Entry
Austria
ContactUsLinkComponent
Anti-cGFP-tag Mouse Monoclonal Antibody
Anti-cGFP-tag Mouse Monoclonal Antibody
Catalog # GENSA00185-40
Supplier:  Genscript
undefined
Anti-cGFP-tag Mouse Monoclonal Antibody
Catalog # GENSA00185-40
Supplier:  Genscript

Specifications

  • Antibody type:
    Tag
  • Antigen symbol:
    cGFP-tag
  • Clonality:
    Monoclonal
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Mouse
  • Isotype:
    IgG2b
  • Western blot:
    Yes
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Size:
    40 µg
  • Pk:
    40 µG

Specifications

About this item

Type: Tag
Antigen: cGFP-tag
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b (Mouse)
Reactivity: