Anti-LSM3 Goat Polyclonal Antibody
EVBIEB11572
- Antibody type:Primary
- Antigen name:LSM3 homolog, U6 small nuclear RNA associated (S. cerevisiae)
- Antigen symbol:LSM3
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Goat
- Isotype:IgG
- Epitope:internal
- Cross adsorption:none
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µg
- Pk:0,1 mg
Type: Primary
Antigen: LSM3 (LSM3 homolog, U6 small nuclear RNA associated (S. cerevisiae))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: internal
Host: Goat
Isotype: IgG
Reactivity: