Anti-ELTD1 Goat polyclonal antibody unconjugated
EVBIEB11420
- Antibody type:Primary
- Antigen name:EGF, latrophilin and seven transmembrane domain containing 1
- Antigen symbol:ELTD1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Goat
- Isotype:IgG
- Reactivity:Zebrafish
- Epitope:internal
- Cross adsorption:none
- Amino acid number:393 to 407
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µg
- Pk:0,1 mg
Type: Primary
Antigen: ELTD1 (EGF, latrophilin and seven transmembrane domain containing 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: internal
Host: Goat
Isotype: IgG
Reactivity: Zebrafish