Anti-Kcnc3 / Kv3.3 Goat Polyclonal Antibody
EVBIEB11267
- Antibody type:Primary
- Antigen symbol:KCNC3
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Goat
- Isotype:IgG
- Reactivity:Pig,Rat,Mouse
- Western blot:Yes
- Epitope:internal
- Cross adsorption:none
- Antigen synonyms:Kv3.3
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µg
- Pk:0,1 mg
Type: Primary
Antigen: Kcnc3 / Kv3.3
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: internal
Host: Goat
Isotype: IgG
Reactivity: Mouse, Pig, Rat