Order Entry
Austria
ContactUsLinkComponent
 
Anti-HRH2 Goat Polyclonal Antibody
  EVBIEB11019
Anti-HRH2 Goat Polyclonal Antibody
  EVBIEB11019

 

  • Antibody type:
    Primary
  • Antigen name:
    histamine receptor H2
  • Antigen symbol:
    HRH2
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Goat
  • Isotype:
    IgG
  • Reactivity:
    Rat,
    Mouse
  • Epitope:
    internal
  • Cross adsorption:
    none
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Size:
    100 µg
  • Pk:
    0,1 mg

 

 

Type: Primary
Antigen: HRH2 (histamine receptor H2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: internal
Host: Goat
Isotype: IgG
Reactivity: Mouse, Rat