Anti-COL11A2 Goat Polyclonal Antibody
EVBIEB11018
- Antibody type:Primary
- Antigen name:collagen, type XI, alpha 2
- Antigen symbol:COL11A2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Goat
- Isotype:IgG
- Reactivity:Mouse
- Epitope:internal
- Cross adsorption:none
- Amino acid number:256 to 268
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µg
- Pk:0,1 mg
Type: Primary
Antigen: COL11A2 (collagen, type XI, alpha 2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: internal
Host: Goat
Isotype: IgG
Reactivity: Mouse