Anti-CETP Goat polyclonal antibody unconjugated
EVBIEB09943
- Antibody type:Primary
- Antigen name:Cholesteryl ester transfer protein, plasma
- Antigen symbol:CETP
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Goat
- Isotype:IgG
- Reactivity:Human
- Epitope:N-terminal
- Cross adsorption:none
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µg
- Pk:0,1 mg
Type: Primary
Antigen: CETP (cholesteryl ester transfer protein, plasma)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Goat
Isotype: IgG
Reactivity: Human